Product Details

SNP ID
rs744434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66080511 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTGAAGCTGATGACCAGAGCA[G/T]TTACTGTGTGCCACAAAACTGCCAG
Phenotype
MIM: 607492
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PACS1 PubMed Links

Gene Details

Gene
PACS1
Gene Name
phosphofurin acidic cluster sorting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018026.3 Intron NP_060496.2
XM_011545162.1 Intron XP_011543464.1
XM_011545164.2 Intron XP_011543466.1

View Full Product Details