Product Details

SNP ID
rs566226
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:65278569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGCTGCTGGTTTTTAAACCAGT[G/T]TGTGAAAATTCCTGGAGCTCAGCAC
Phenotype
MIM: 608375
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC6 PubMed Links

Gene Details

Gene
DNAJC6
Gene Name
DnaJ heat shock protein family (Hsp40) member C6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256864.1 Intron NP_001243793.1
NM_001256865.1 Intron NP_001243794.1
NM_014787.3 Intron NP_055602.1

View Full Product Details