Product Details

SNP ID
rs175498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75069666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGCCAGCTGAGAGCGCCTGCGC[A/G]GCTGTCGGTTGATGGGGAGGTCAGG
Phenotype
MIM: 600875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACYP1 PubMed Links

Gene Details

Gene
ACYP1
Gene Name
acylphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001107.4 26 Intron NP_001098.1
NM_001302616.1 26 Intron NP_001289545.1
NM_001302617.1 26 Intron NP_001289546.1
Gene
ZC2HC1C
Gene Name
zinc finger C2HC-type containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042430.2 26 UTR 5 NP_001035895.1
NM_024643.3 26 UTR 5 NP_078919.2
XM_005268062.3 26 Intron XP_005268119.1
XM_005268063.4 26 UTR 5 XP_005268120.1
XM_006720255.3 26 UTR 5 XP_006720318.1
XM_011537155.2 26 Intron XP_011535457.1
XM_011537156.2 26 UTR 5 XP_011535458.1
XM_011537157.2 26 UTR 5 XP_011535459.1
XM_017021643.1 26 UTR 5 XP_016877132.1
XM_017021644.1 26 Intron XP_016877133.1
XM_017021645.1 26 UTR 5 XP_016877134.1

View Full Product Details