Product Details

SNP ID
rs729751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6945187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGGATTTTCTAATTAAGGATGG[C/T]CAACCTGTACTGTTAATCACATTAT
Phenotype
MIM: 607462 MIM: 615140 MIM: 176883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATN1 PubMed Links

Gene Details

Gene
ATN1
Gene Name
atrophin 1
There are no transcripts associated with this gene.

Gene
C12orf57
Gene Name
chromosome 12 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301834.1 Intron NP_001288763.1
NM_001301836.1 Intron NP_001288765.1
NM_001301837.1 Intron NP_001288766.1
NM_001301838.1 Intron NP_001288767.1
NM_138425.3 Intron NP_612434.1
Gene
PTPN6
Gene Name
protein tyrosine phosphatase, non-receptor type 6
There are no transcripts associated with this gene.

Gene
RNU7-1
Gene Name
RNA, U7 small nuclear 1
There are no transcripts associated with this gene.

View Full Product Details