Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318245.1 | Intron | NP_001305174.1 | |||
NM_032608.6 | Intron | NP_115997.5 | |||
XM_011530458.2 | Intron | XP_011528760.1 | |||
XM_011530459.2 | Intron | XP_011528761.1 | |||
XM_011530460.2 | Intron | XP_011528762.1 | |||
XM_011530461.2 | Intron | XP_011528763.1 | |||
XM_011530464.2 | Intron | XP_011528766.1 | |||
XM_011530465.2 | Intron | XP_011528767.1 | |||
XM_011530466.2 | Intron | XP_011528768.1 | |||
XM_017029012.1 | Intron | XP_016884501.1 | |||
XM_017029013.1 | Intron | XP_016884502.1 | |||
XM_017029014.1 | Intron | XP_016884503.1 | |||
XM_017029015.1 | Intron | XP_016884504.1 | |||
XM_017029016.1 | Intron | XP_016884505.1 | |||
XM_017029017.1 | Intron | XP_016884506.1 |