Product Details

SNP ID
rs395917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45225829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCACGGCGCCGCCGCCGCCGCCG[C/T]CTCCTCCGGGCCCGGGCGCTCGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM158 PubMed Links

Gene Details

Gene
TMEM158
Gene Name
transmembrane protein 158 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015444.2 494 Missense Mutation ACG,GCG T67A NP_056259.2

View Full Product Details