Product Details

SNP ID
rs176880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:38112489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATTAAAAAAAATTTGTGCAAC[C/G]TCAGACATAAACGGGCTATAATGTA
Phenotype
MIM: 616085
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF37A PubMed Links

Gene Details

Gene
ZNF37A
Gene Name
zinc finger protein 37A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007094.2 Intron NP_001007095.1
NM_001178101.1 Intron NP_001171572.1
NM_001324245.1 Intron NP_001311174.1
NM_001324246.1 Intron NP_001311175.1
NM_001324247.1 Intron NP_001311176.1
NM_001324248.1 Intron NP_001311177.1
NM_001324249.1 Intron NP_001311178.1
NM_001324250.1 Intron NP_001311179.1
NM_001324251.1 Intron NP_001311180.1
NM_001324252.1 Intron NP_001311181.1
NM_001324253.1 Intron NP_001311182.1
NM_001324254.1 Intron NP_001311183.1
NM_001324256.1 Intron NP_001311185.1
NM_001324257.1 Intron NP_001311186.1
NM_001324258.1 Intron NP_001311187.1
NM_001324259.1 Intron NP_001311188.1
NM_001324260.1 Intron NP_001311189.1
NM_001324261.1 Intron NP_001311190.1
NM_001324262.1 Intron NP_001311191.1
NM_003421.2 Intron NP_003412.1
XM_005252586.3 Intron XP_005252643.1
XM_011519656.2 Intron XP_011517958.1
XM_011519657.2 Intron XP_011517959.1
XM_011519658.2 Intron XP_011517960.1
XM_017016618.1 Intron XP_016872107.1
XM_017016619.1 Intron XP_016872108.1
XM_017016620.1 Intron XP_016872109.1
XM_017016621.1 Intron XP_016872110.1
XM_017016622.1 Intron XP_016872111.1
XM_017016623.1 Intron XP_016872112.1
XM_017016624.1 Intron XP_016872113.1

View Full Product Details