Product Details

SNP ID
rs1051500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58433035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCACAGTCCTGAATCCCTAGAG[A/G]ACACAGGAAGGAAGGCTCAGGGTAC
Phenotype
MIM: 604074
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF132 PubMed Links

Gene Details

Gene
ZNF132
Gene Name
zinc finger protein 132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003433.3 2810 UTR 3 NP_003424.3

View Full Product Details