Product Details

SNP ID
rs1110331
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:89707988 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGGCAGGGGCAGCCGGTGTCATCT[G/A]AACGAACAGAAGGATACCCCCGGCC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SPATA2L PubMed Links

Gene Details

Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

Gene
VPS9D1
Gene Name
VPS9 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004913.2 Intron NP_004904.2
XM_005256329.4 Intron XP_005256386.1
XM_005256330.4 Intron XP_005256387.1
XM_005256331.2 Intron XP_005256388.1
XM_006721350.2 Intron XP_006721413.1
XM_011523476.2 Intron XP_011521778.1
XM_011523477.2 Intron XP_011521779.1
XM_011523478.2 Intron XP_011521780.1
XM_011523479.2 Intron XP_011521781.1
XM_011523480.2 Intron XP_011521782.1
Gene
VPS9D1-AS1
Gene Name
VPS9D1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details