Product Details

SNP ID
rs10433890
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:98878771 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCAACTGCCAAAAGTGATTCAGC[A/G]TAATTTATTATCCTTTCTTGTCCCC
Phenotype
MIM: 133440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4E PubMed Links

Gene Details

Gene
EIF4E
Gene Name
eukaryotic translation initiation factor 4E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130678.1 4434 UTR 3 NP_001124150.1
NM_001130679.1 4434 UTR 3 NP_001124151.1
NM_001968.3 4434 UTR 3 NP_001959.1
XM_006714126.3 4434 Intron XP_006714189.1

View Full Product Details