Product Details

SNP ID
rs8102871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37915829 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGCACTTTGATTTGCTGTAGC[C/G]TGGGCGTATTTTGAATGAAGGGTAG
Phenotype
MIM: 616655
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SIPA1L3 PubMed Links

Gene Details

Gene
SIPA1L3
Gene Name
signal induced proliferation associated 1 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015073.2 Intron NP_055888.1
XM_005258671.4 Intron XP_005258728.1
XM_011526657.2 Intron XP_011524959.1
XM_017026517.1 Intron XP_016882006.1
XM_017026518.1 Intron XP_016882007.1
Gene
WDR87
Gene Name
WD repeat domain 87
There are no transcripts associated with this gene.

View Full Product Details