Product Details

SNP ID
rs12611436
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222681328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGGCTACAAATTGAGGATTCACAC[A/G]ACCTCCTCCTTGGACTCTGTCATTT
Phenotype
MIM: 610268
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MOGAT1 PubMed Links

Gene Details

Gene
MOGAT1
Gene Name
monoacylglycerol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058165.2 Intron NP_477513.2

View Full Product Details