Product Details

SNP ID
rs9508485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19553745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACGTACAATTCTAAAAGATGCAA[A/G]CTAATCTATTGTAGATCAATGGTTG
Phenotype
MIM: 606791
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPTE2 PubMed Links

Gene Details

Gene
TPTE2
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141968.1 Intron NP_001135440.1
NM_001271850.1 Intron NP_001258779.1
NM_130785.3 Intron NP_570141.3
NM_199254.2 Intron NP_954863.2

View Full Product Details