Product Details

SNP ID
rs1105881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41780332 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGGCTTCAGCCTGACATCTGTAA[C/G]CTCCCCACCAACCCTCTGAGTCTGA
Phenotype
MIM: 616786
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAPKBP1 PubMed Links

Gene Details

Gene
MAPKBP1
Gene Name
mitogen-activated protein kinase binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128608.1 Intron NP_001122080.1
NM_001265611.1 Intron NP_001252540.1
NM_014994.2 Intron NP_055809.2
XM_006720438.1 Intron XP_006720501.1
XM_006720439.2 Intron XP_006720502.1
XM_011521383.1 Intron XP_011519685.1
XM_011521384.2 Intron XP_011519686.1
XM_017022017.1 Intron XP_016877506.1

View Full Product Details