Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001085488.2 | Intron | NP_001078957.1 | |||
NM_001282011.1 | Intron | NP_001268940.1 | |||
XM_005258812.3 | Intron | XP_005258869.1 | |||
XM_011526850.2 | Intron | XP_011525152.1 | |||
XM_011526851.2 | Intron | XP_011525153.1 | |||
XM_011526852.2 | Intron | XP_011525154.1 | |||
XM_011526853.2 | Intron | XP_011525155.1 | |||
XM_011526854.2 | Intron | XP_011525156.1 | |||
XM_011526855.2 | Intron | XP_011525157.1 | |||
XM_011526856.2 | Intron | XP_011525158.1 | |||
XM_011526857.2 | Intron | XP_011525159.1 | |||
XM_017026667.1 | Intron | XP_016882156.1 | |||
XM_017026668.1 | Intron | XP_016882157.1 |