Product Details

SNP ID
rs12679208
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:73304015 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATTTCTAACCTAAATGAAAAAAAC[A/T]GCAATGCTCCCCTCATTGCCAAATC
Phenotype
MIM: 607599 MIM: 604166
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RDH10 PubMed Links

Gene Details

Gene
RDH10
Gene Name
retinol dehydrogenase 10 (all-trans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172037.4 Intron NP_742034.1
Gene
RDH10-AS1
Gene Name
RDH10 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPL7
Gene Name
ribosomal protein L7
There are no transcripts associated with this gene.

View Full Product Details