Product Details

SNP ID
rs8073626
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78794197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCCACTTCTCAGCCAACCTGG[C/T]GGCTGGTCATGCTCACCTGCTGTTC
Phenotype
MIM: 612543
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
USP36 PubMed Links

Gene Details

Gene
USP36
Gene Name
ubiquitin specific peptidase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321291.1 Intron NP_001308220.1
XM_005257542.2 Intron XP_005257599.1
XM_005257543.2 Intron XP_005257600.1
XM_011525065.1 Intron XP_011523367.1
XM_011525066.2 Intron XP_011523368.1
XM_011525067.2 Intron XP_011523369.1
XM_011525068.1 Intron XP_011523370.1
XM_011525069.2 Intron XP_011523371.1
XM_011525070.1 Intron XP_011523372.1
XM_011525071.2 Intron XP_011523373.1
XM_017024900.1 Intron XP_016880389.1
XM_017024901.1 Intron XP_016880390.1

View Full Product Details