Product Details

SNP ID
rs12986332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38733450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTCCTGGAGGGGCCTCTCCCT[C/G]CCCCAGAACCCCTACCAGGCACATC
Phenotype
MIM: 604638 MIM: 608839
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACTN4 PubMed Links

Gene Details

Gene
ACTN4
Gene Name
actinin alpha 4
There are no transcripts associated with this gene.

Gene
CAPN12
Gene Name
calpain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144691.4 Intron NP_653292.2
XM_017026355.1 Intron XP_016881844.1
XM_017026356.1 Intron XP_016881845.1
XM_017026357.1 Intron XP_016881846.1
XM_017026358.1 Intron XP_016881847.1
XM_017026359.1 Intron XP_016881848.1
XM_017026360.1 Intron XP_016881849.1
XM_017026361.1 Intron XP_016881850.1
XM_017026362.1 Intron XP_016881851.1
XM_017026363.1 Intron XP_016881852.1
XM_017026364.1 Intron XP_016881853.1
XM_017026365.1 Intron XP_016881854.1

View Full Product Details