Product Details
- SNP ID
-
rs4802770
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:38732762 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGCCCCCATCTCCCTGTGGCTCC[A/C]TCTGCTGTGTGGTGGCAGACAGGCC
- Phenotype
-
MIM: 604638
MIM: 608839
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTN4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138146294] are located under a probe and SNP(s) [rs141454785] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACTN4
- Gene Name
- actinin alpha 4
There are no transcripts associated with this gene.
- Gene
- CAPN12
- Gene Name
- calpain 12
View Full Product Details