Product Details
- SNP ID
-
rs12521445
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150934694 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATACACAGGATTTTACATTTTTT[A/T]AAATATGCAGGTGCTACAGTTTGAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF300P1
PubMed Links
Gene Details
- Gene
- ZNF300P1
- Gene Name
- zinc finger protein 300 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details