Product Details

SNP ID
rs6877568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:31532578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCATCGGAGGCCAGAGAGTTAA[A/C]CAGAGTTAAACTGCCCTATTCCGTT
Phenotype
MIM: 608828
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C5orf22 PubMed Links

Gene Details

Gene
C5orf22
Gene Name
chromosome 5 open reading frame 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018356.2 Intron NP_060826.2
XM_006714479.2 Intron XP_006714542.1
XM_006714480.3 Intron XP_006714543.1
XM_011514062.2 Intron XP_011512364.1
XM_017009607.1 Intron XP_016865096.1
XM_017009608.1 Intron XP_016865097.1
XM_017009609.1 Intron XP_016865098.1
XM_017009610.1 Intron XP_016865099.1
XM_017009611.1 Intron XP_016865100.1
XM_017009612.1 Intron XP_016865101.1
XM_017009613.1 Intron XP_016865102.1
XM_017009614.1 Intron XP_016865103.1
XM_017009615.1 Intron XP_016865104.1
XM_017009616.1 Intron XP_016865105.1
Gene
DROSHA
Gene Name
drosha ribonuclease III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100412.1 Intron NP_001093882.1
NM_013235.4 Intron NP_037367.3
XM_005248291.3 Intron XP_005248348.1
XM_005248292.3 Intron XP_005248349.1
XM_005248293.3 Intron XP_005248350.1
XM_005248294.3 Intron XP_005248351.1
XM_011514033.2 Intron XP_011512335.1
XM_017009399.1 Intron XP_016864888.1
XM_017009400.1 Intron XP_016864889.1
XM_017009401.1 Intron XP_016864890.1
XM_017009402.1 Intron XP_016864891.1

View Full Product Details