Product Details

SNP ID
rs1146297
Assay Type
Functionally tested
NCBI dbSNP Submissions
72
Location
Chr.1:117368060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGTGCCCTCTTAAAAAGCACAACA[A/G]TCCTTTAAGAGGAGCAAAATTGAGT
Phenotype
MIM: 604345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAN1A2 PubMed Links

Gene Details

Gene
MAN1A2
Gene Name
mannosidase alpha class 1A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006699.3 606 UTR 5 NP_006690.1
XM_006710302.2 606 UTR 5 XP_006710365.1
XM_011540536.2 606 UTR 5 XP_011538838.1
XM_017000115.1 606 UTR 5 XP_016855604.1

View Full Product Details