Product Details

SNP ID
rs7210207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35409795 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCCATCTTGAGGCTGCTGTTTCT[A/G]CATTCATCCTCCCATGGTCACATTT
Phenotype
MIM: 614955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLFN12 PubMed Links

Gene Details

Gene
SLFN12
Gene Name
schlafen family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289009.1 3548 Intron NP_001275938.1
NM_018042.4 3548 Intron NP_060512.3
XM_005257995.4 3548 UTR 3 XP_005258052.1
XM_011524967.2 3548 UTR 3 XP_011523269.1
XM_017024809.1 3548 UTR 3 XP_016880298.1
XM_017024810.1 3548 UTR 3 XP_016880299.1
XM_017024811.1 3548 Intron XP_016880300.1

View Full Product Details