Product Details
- SNP ID
-
rs35092254
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:155672581 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGCTATTAAAAACAAAAGGAGATAA[C/G]TAGGTTAGTTGACAAGGATAGGTTC
- Phenotype
-
MIM: 139396
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GUCY1A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73858154] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GUCY1A3
- Gene Name
- guanylate cyclase 1 soluble subunit alpha
View Full Product Details