Product Details

SNP ID
rs9085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20685213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGGAGTAACATTTAAAATAACAG[A/C]TAAAAGAAAAGCTCTGAATGTTACT
Phenotype
MIM: 613570
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LDAH PubMed Links

Gene Details

Gene
LDAH
Gene Name
lipid droplet associated hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282719.1 2620 UTR 3 NP_001269648.1
NM_001282720.1 2620 UTR 3 NP_001269649.1
NM_001282721.1 2620 UTR 3 NP_001269650.1
NM_001282722.1 2620 UTR 3 NP_001269651.1
NM_001282723.1 2620 UTR 3 NP_001269652.1
NM_001282724.1 2620 UTR 3 NP_001269653.1
NM_021925.3 2620 UTR 3 NP_068744.1
XM_011533014.2 2620 UTR 3 XP_011531316.1

View Full Product Details