Product Details
- SNP ID
-
rs1045926
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:56620572 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCATGTACCAATACCCTCTGCATTT[C/G]AAAATGTTGACTCAGATGTTTTTCC
- Phenotype
-
MIM: 616493
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC66
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs534538641] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC66
- Gene Name
- coiled-coil domain containing 66
- Gene
- FAM208A
- Gene Name
- family with sequence similarity 208 member A
View Full Product Details