Product Details
- SNP ID
-
rs34362405
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:93724000 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCAAGCACTAAAATTACACTGGT[A/G]TTTACCCTTTGTCCACAAACAGAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSG1
PubMed Links
Gene Details
- Gene
- TSG1
- Gene Name
- tumor suppressor TSG1
There are no transcripts associated with this gene.
View Full Product Details