Product Details

SNP ID
rs13315088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149967351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGTTTGACGCTTCTTTTCAGC[A/G]TATAGTGCAGCTAAGGTTTAAAGAG
Phenotype
MIM: 176590 MIM: 609247
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC646903 PubMed Links

Gene Details

Gene
LOC646903
Gene Name
uncharacterized LOC646903
There are no transcripts associated with this gene.

Gene
PFN2
Gene Name
profilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002628.4 Intron NP_002619.1
NM_053024.3 Intron NP_444252.1
Gene
RNF13
Gene Name
ring finger protein 13
There are no transcripts associated with this gene.

View Full Product Details