Product Details

SNP ID
rs1486961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:140231944 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATACAACTTAAATAAATCGCACTC[A/G]TTTCAACTCAAATTTGAAGATAAAC
Phenotype
MIM: 608766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP1B PubMed Links

Gene Details

Gene
LRP1B
Gene Name
LDL receptor related protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017004342.1 10013 UTR 3 XP_016859831.1

View Full Product Details