Product Details

SNP ID
rs1490496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184117056 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATAAATCTTTGAATATATAGACT[C/T]TCAAAATCAGAAGAAGCTTTCAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSP90AA5P PubMed Links

Gene Details

Gene
HSP90AA5P
Gene Name
heat shock protein 90 alpha family class A member 5, pseudogene
There are no transcripts associated with this gene.

View Full Product Details