Product Details

SNP ID
rs7654770
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:48487250 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATACGTAAGCATCATTACAGACTTA[C/T]AGATGGGAGGTTTCTCATGAACAAA
Phenotype
MIM: 607520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC10A4 PubMed Links

Gene Details

Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152679.3 Intron NP_689892.1
Gene
ZAR1
Gene Name
zygote arrest 1
There are no transcripts associated with this gene.

View Full Product Details