Product Details

SNP ID
rs9825882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100187406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTGTTGACTATGCAGATGAGT[A/G]AGATGAGGAAGAGTGGATTTGTGCC
Phenotype
MIM: 611030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMSS1 PubMed Links

Gene Details

Gene
CMSS1
Gene Name
cms1 ribosomal small subunit homolog (yeast)
There are no transcripts associated with this gene.

Gene
TMEM30C
Gene Name
transmembrane protein 30C
There are no transcripts associated with this gene.

View Full Product Details