Product Details

SNP ID
rs11733884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70402026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAACAGAACATAAATAAAATAC[G/T]TTACAGAAAGGGCTATGATAAAATA
Phenotype
MIM: 608936
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OPRPN PubMed Links

Gene Details

Gene
OPRPN
Gene Name
opiorphin prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302807.1 Intron NP_001289736.1
NM_021225.4 Intron NP_067048.4

View Full Product Details