Product Details

SNP ID
rs17649675
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:75920486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGGTAGGTAAAATAGACACATCC[C/T]CTGTGCTTACAAGACTCACGGTGTA
Phenotype
MIM: 602105
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB17 PubMed Links

Gene Details

Gene
ASB17
Gene Name
ankyrin repeat and SOCS box containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080868.2 Intron NP_543144.1
XM_006710345.3 Intron XP_006710408.1
XM_006710346.3 Intron XP_006710409.1
Gene
MSH4
Gene Name
mutS homolog 4
There are no transcripts associated with this gene.

View Full Product Details