Product Details

SNP ID
rs518770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39938071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACCCTCACACCCCAAGTCGTTTG[C/T]TAAGGAGAGCACCACGTCATCTCTT
Phenotype
MIM: 609280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2AK4 PubMed Links

Gene Details

Gene
EIF2AK4
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013703.3 Intron NP_001013725.2
XM_005254392.2 Intron XP_005254449.1
XM_011521599.2 Intron XP_011519901.1
XM_011521600.2 Intron XP_011519902.1
XM_017022219.1 Intron XP_016877708.1

View Full Product Details