Product Details

SNP ID
rs6414900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43126426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCCATGCCATTGTGCAGTTCTCA[A/G]CGCCTTTCCTCTGCCGCCTTGGGTC
Phenotype
MIM: 604073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF131 PubMed Links

Gene Details

Gene
ZNF131
Gene Name
zinc finger protein 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005248359.2 Intron XP_005248416.1
XM_005248360.2 Intron XP_005248417.1
XM_005248361.2 Intron XP_005248418.1
XM_005248362.4 Intron XP_005248419.1
XM_005248363.4 Intron XP_005248420.1
XM_005248365.3 Intron XP_005248422.1
XM_005248367.2 Intron XP_005248424.1
XM_011514123.2 Intron XP_011512425.1
XM_011514124.2 Intron XP_011512426.1
XM_017009830.1 Intron XP_016865319.1
XM_017009831.1 Intron XP_016865320.1
XM_017009832.1 Intron XP_016865321.1
XM_017009833.1 Intron XP_016865322.1
XM_017009834.1 Intron XP_016865323.1
XM_017009835.1 Intron XP_016865324.1
XM_017009836.1 Intron XP_016865325.1
XM_017009837.1 Intron XP_016865326.1
XM_017009838.1 Intron XP_016865327.1
XM_017009839.1 Intron XP_016865328.1
XM_017009840.1 Intron XP_016865329.1
XM_017009841.1 Intron XP_016865330.1
XM_017009842.1 Intron XP_016865331.1
XM_017009843.1 Intron XP_016865332.1

View Full Product Details