Product Details

SNP ID
rs10070923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:172200387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCAAGTTCCTTTTGAATGCAGA[C/T]GCTGAAGTGGGACTTGCTGCATTTG
Phenotype
MIM: 610174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFCAB9 PubMed Links

Gene Details

Gene
EFCAB9
Gene Name
EF-hand calcium binding domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171183.1 Intron NP_001164654.1
XM_017009350.1 Intron XP_016864839.1
Gene
UBTD2
Gene Name
ubiquitin domain containing 2
There are no transcripts associated with this gene.

View Full Product Details