Product Details

SNP ID
rs2930945
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:69218876 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGAATGATCCTTACTTATTCAGT[A/G]GGTAGTTACTGAGCATCAACTAGTT
Phenotype
MIM: 605607 MIM: 611996
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPH PubMed Links

Gene Details

Gene
CENPH
Gene Name
centromere protein H
There are no transcripts associated with this gene.

Gene
MRPS36
Gene Name
mitochondrial ribosomal protein S36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033281.5 Intron NP_150597.1

View Full Product Details