Product Details

SNP ID
rs1994719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74949799 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACCCCTCTCCAGCCAGGACTCAG[A/G]GGTCTGCCCCCTGCTGCGAGAAGAG
Phenotype
MIM: 605752 MIM: 607828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HID1 PubMed Links

Gene Details

Gene
HID1
Gene Name
HID1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030630.2 2240 Intron NP_085133.1
XM_005257226.1 2240 Intron XP_005257283.1
Gene
OTOP3
Gene Name
otopetrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272005.1 2240 UTR 3 NP_001258934.1
NM_178233.2 2240 UTR 3 NP_839947.1
XM_011524744.2 2240 UTR 3 XP_011523046.1

View Full Product Details