Product Details

SNP ID
rs12418753
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:7020532 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCGGGGGATAAGGCGAGGACGCAC[C/T]AGCATTAATGGAATAGGAGAACTCC
Phenotype
MIM: 609665 MIM: 605015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLRP14 PubMed Links

Gene Details

Gene
NLRP14
Gene Name
NLR family pyrin domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176822.3 64 UTR 5 NP_789792.1
XM_011520044.1 64 UTR 5 XP_011518346.1
Gene
ZNF214
Gene Name
zinc finger protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013249.2 64 Intron NP_037381.2
XM_005253128.3 64 Intron XP_005253185.1
XM_005253129.3 64 Intron XP_005253186.1
XM_006718308.3 64 Intron XP_006718371.1
XM_006718309.3 64 Intron XP_006718372.1
XM_011520355.2 64 Intron XP_011518657.1
XM_011520357.2 64 Intron XP_011518659.1
XM_017018269.1 64 Intron XP_016873758.1

View Full Product Details