Product Details

SNP ID
rs11813395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70829713 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAGTGAGAGCTAAAATTAGAGCC[A/G]AAGGTTTTGATTACCAGTCCAGACT
Phenotype
MIM: 603729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGPL1 PubMed Links

Gene Details

Gene
SGPL1
Gene Name
sphingosine-1-phosphate lyase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003901.3 Intron NP_003892.2
XM_005270263.1 Intron XP_005270320.1
XM_006718053.1 Intron XP_006718116.1
XM_011540316.2 Intron XP_011538618.1
XM_011540317.1 Intron XP_011538619.1
XM_011540318.1 Intron XP_011538620.1

View Full Product Details