Product Details

SNP ID
rs9985834
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:67823614 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTATTATTACCAAGTGTGCTTCTAA[A/G]TTAGTCAGCTATTCAGTTGGCTGCT
Phenotype
MIM: 605369
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11D PubMed Links

Gene Details

Gene
TMPRSS11D
Gene Name
transmembrane protease, serine 11D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004262.2 Intron NP_004253.1
XM_005265710.4 Intron XP_005265767.1
XM_017008851.1 Intron XP_016864340.1

View Full Product Details