Product Details

SNP ID
rs13387185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:67401517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTTATAATGAAATGAAGTAAT[G/T]CATTTAGATACCTTATACAGCACCT
Phenotype
MIM: 613196
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ETAA1 PubMed Links

Gene Details

Gene
ETAA1
Gene Name
Ewing tumor associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019002.3 Intron NP_061875.2
XM_017004376.1 Intron XP_016859865.1
XM_017004377.1 Intron XP_016859866.1

View Full Product Details