Product Details

SNP ID
rs1083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1422003 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTGACCCCATATAAATCATTCC[A/G]TACTTAAATGGGTTTGTAAACTAAA
Phenotype
MIM: 164762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005206.4 2575 UTR 3 NP_005197.3
NM_016823.3 2575 UTR 3 NP_058431.2

View Full Product Details