Product Details

SNP ID
rs423475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52979785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGTAACAAAAACCCTAATAAT[G/T]ATGCCAATTAATTATATATAAATAT
Phenotype
MIM: 605450
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GSTA4 PubMed Links

Gene Details

Gene
GSTA4
Gene Name
glutathione S-transferase alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001512.3 Intron NP_001503.1
XM_005249035.3 Intron XP_005249092.1
XM_011514534.2 Intron XP_011512836.1
XM_011514535.2 Intron XP_011512837.1

View Full Product Details