Product Details

SNP ID
rs9432382
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:94822131 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGGCCTGCCTTGTGCCTGGCAG[C/T]TTTGACCTCAAGCTGTTATTGTGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01057 PubMed Links

Gene Details

Gene
LINC01057
Gene Name
long intergenic non-protein coding RNA 1057
There are no transcripts associated with this gene.

Gene
SLC44A3
Gene Name
solute carrier family 44 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114106.2 Intron NP_001107578.1
NM_001258340.1 Intron NP_001245269.1
NM_001258341.1 Intron NP_001245270.1
NM_001258342.1 Intron NP_001245271.1
NM_001258343.1 Intron NP_001245272.1
NM_001301079.1 Intron NP_001288008.1
NM_152369.4 Intron NP_689582.2
XM_005270440.2 Intron XP_005270497.1
XM_005270441.2 Intron XP_005270498.1
XM_005270442.1 Intron XP_005270499.1
XM_011540632.2 Intron XP_011538934.1
XM_011540633.2 Intron XP_011538935.1
XM_011540635.2 Intron XP_011538937.1
XM_011540636.2 Intron XP_011538938.1
XM_017000262.1 Intron XP_016855751.1
XM_017000263.1 Intron XP_016855752.1
XM_017000264.1 Intron XP_016855753.1
XM_017000265.1 Intron XP_016855754.1
XM_017000266.1 Intron XP_016855755.1

View Full Product Details