Product Details

SNP ID
rs12039319
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:100874578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTACATTTCAGTTTTATCTAGCTC[C/T]AGGTAACTTCATAAATCTGTGTTCT
Phenotype
MIM: 602411
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXTL2 PubMed Links

Gene Details

Gene
EXTL2
Gene Name
exostosin like glycosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033025.2 Intron NP_001028197.1
NM_001261440.1 Intron NP_001248369.1
NM_001261441.1 Intron NP_001248370.1
NM_001261442.1 Intron NP_001248371.1
NM_001439.3 Intron NP_001430.1
XM_005270621.1 Intron XP_005270678.1
XM_011540994.2 Intron XP_011539296.1
XM_011540995.2 Intron XP_011539297.1
XM_011540996.2 Intron XP_011539298.1
XM_017000651.1 Intron XP_016856140.1

View Full Product Details