Product Details

SNP ID
rs3795663
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:119622564 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CTTACTCAACACTCCTCCCACTTCA[A/G]GAAACCCCAAACACCAAAGGCTCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF697 PubMed Links

Gene Details

Gene
ZNF697
Gene Name
zinc finger protein 697
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080470.1 4898 UTR 3 NP_001073939.1
XM_005271315.3 4898 UTR 3 XP_005271372.1
XM_011542416.2 4898 UTR 3 XP_011540718.1
XM_017002783.1 4898 UTR 3 XP_016858272.1

View Full Product Details