Product Details

SNP ID
rs11698289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44308843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGATATGAGCCATGCACCTGGCCT[C/G]GTTCTTTCCTAATGGCAGAAACAGT
Phenotype
MIM: 612029
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FITM2 PubMed Links

Gene Details

Gene
FITM2
Gene Name
fat storage inducing transmembrane protein 2
There are no transcripts associated with this gene.

View Full Product Details