Product Details
- SNP ID
-
rs6837966
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:7101752 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GCTCTAAGAATCTGGGGATGAAACA[C/G]GCTGGGCAGGGAGGGGTCCTTTGGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ36777
PubMed Links
Gene Details
- Gene
- FLJ36777
- Gene Name
- uncharacterized LOC730971
There are no transcripts associated with this gene.
View Full Product Details